missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human IGFBP-5 Biotinylated Antibody, R&D Systems™

Goat Polyclonal Antibody
Brand: R&D Systems BAF875
This item is not returnable.
View return policy
Description
IGFBP-5 Polyclonal specifically detects IGFBP-5 in Human samples. It is validated for Western Blot, ELISA Detection (Matched Antibody Pair).
Specifications
| IGFBP-5 | |
| Polyclonal | |
| Biotin | |
| P24593 | |
| IGFBP5 | |
| Mouse myeloma cell line NS0-derived recombinant human IGFBP-5 (R&D Systems, Catalog # 875-B5) Glu28-Glu272 Accession # P24593 | |
| 50 μg | |
| Primary | |
| Detects human IGFBP-5 in ELISAs and Western blots. In sandwich immunoassays, less than 10% cross-reactivity with recombinant mouse IGFBP-5 is observed and less than 0.5% cross-reactivity with recombinant human (rh) IGFBP-2, rhIGFBP-3, rhIGFBP-4, and rhIGFBP-6 is observed. | |
| Human | |
| IgG |
| Western Blot, ELISA | |
| LYOPH | |
| Western Blot 0.1 ug/mL, ELISA Detection (Matched Antibody Pair) 0.1-0.4 ug/mL | |
| IBP5IBP-5, IGF-binding protein 5, IGFBP5, IGFBP-5, insulin-like growth factor binding protein 5, insulin-like growth factor-binding protein 5 | |
| Goat | |
| Affinity Purified | |
| RUO | |
| 3488 | |
| Reconstitute at 0.2 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
Berichtigung von Produktinhalten
Bitte geben Sie uns Ihr Feedback zu den Produktinhalten, indem Sie das folgende Formular ausfüllen.
Name des Produkts
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur