missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HSFY1 Full-length ORF (NP_149099.2, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00086614-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16116373

  • 508.00€ / 25µg
Estimated Shipment: 13-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]

Sequence: MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYN
Specifications

Specifications

NP_149099.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
71.5kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HSF2L/HSFY
HSFY1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
86614
HSFY1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYN
RUO
HSFY1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers