missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human HSFY1 Full-length ORF (NP_149099.2, 1 a.a. - 401 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_149099.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 86614 |
Molecular Weight (g/mol) | 71.5kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16116373
|
Abnova™
H00086614-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16106373
|
Abnova™
H00086614-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the heat shock factor (HSF) family of transcriptional activators for heat shock proteins. This gene is a candidate gene for azoospermia, since it localizes to a region of chromosome Y that is sometimes deleted in infertile males. The genome has two identical copies of this gene within a palindromic region; this record represents the more centromeric copy. Alternative splicing results in multiple transcript variants encoding distinct isoforms. [provided by RefSeq]
Sequence: MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYNSpecifications
NP_149099.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
71.5kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAHVSSETQDVSPKDELTASEASTRSPLCEHTFPGDSDLRSMIEEHAFQVLSQGSLLESPSYTVCVSEPDKDDDFLSLNFPRKLWKIVESDQFKSISWDENGTCIVINEELFKKEILETKAPYRIFQTDAIKSFVRQLNLYGFSKIQQNFQRSAFLATFLSEEKESSVLSKLKFYYNPNFKRGYPQLLVRVKRRIGVKNASPISTLFNEDFNKKHFRAGANMENHNSALAAEASEESLFSASKNLNMPLTRESSVRQIIANSSVPIRSGFPPPSPSTSVGPSEQIATDQHAILNQLTTIHMHSHSTYMQARGHIVNFITTTTSQYHIISPLQNGYFGLTVEPSAVPTRYPLVSVNEAPYRNMLPAGNPWLQMPTIADRSAAPHSRLALQPSPLDKYHPNYN | |
RUO | |
HSFY1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
86614 | |
HSFY1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HSF2L/HSFY | |
HSFY1 | |
Recombinant | |
wheat germ expression system |