Learn More
Abnova™ Human HOXB6 Partial ORF (NP_724779.1, 1 a.a. - 57 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_724779.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 3216 |
Molecular Weight (g/mol) | 32.01kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16024165
|
Abnova™
H00003216-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
16014165
|
Abnova™
H00003216-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-06-2024
Log in to see stock. |
|||||
Description
This gene is a member of the Antp homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development, including that of lung and skin, and has been localized to both the nucleus and cytoplasm. Altered expression of this gene or a change in the subcellular localization of its protein is associated with some cases of acute myeloid leukemia and colorectal cancer. [provided by RefSeq]
Sequence: MSSYFVNSTFPVTLASGQESFLGQLPLYSSGYADPLRHYPAPYGPGPGQDKGFATSSSpecifications
NP_724779.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.01kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX2/HOX2B/HU-2/Hox-2.2 | |
HOXB6 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
3216 | |
HOXB6 (Human) Recombinant Protein (Q01) | |
MSSYFVNSTFPVTLASGQESFLGQLPLYSSGYADPLRHYPAPYGPGPGQDKGFATSS | |
RUO | |
HOXB6 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |