missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human HOXB6 Partial ORF (NP_724779.1, 1 a.a. - 57 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00003216-Q01.25ug

Additional Details : Weight : 0.02000kg

 View more versions of this product

Product Code. 16024165

  • 508.00€ / 25µg
Estimated Shipment:
to see stock.
Add to basket

Description

Description

This gene is a member of the Antp homeobox family and encodes a protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development, including that of lung and skin, and has been localized to both the nucleus and cytoplasm. Altered expression of this gene or a change in the subcellular localization of its protein is associated with some cases of acute myeloid leukemia and colorectal cancer. [provided by RefSeq]

Sequence: MSSYFVNSTFPVTLASGQESFLGQLPLYSSGYADPLRHYPAPYGPGPGQDKGFATSS
Specifications

Specifications

NP_724779.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
32.01kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
MSSYFVNSTFPVTLASGQESFLGQLPLYSSGYADPLRHYPAPYGPGPGQDKGFATSS
RUO
HOXB6
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, ELISA, Protein Array, Western Blot
3216
HOXB6 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HOX2/HOX2B/HU-2/Hox-2.2
HOXB6
Recombinant
wheat germ expression system
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers