Learn More
Abnova™ Human HOXB4 Partial ORF (NP_076920, 1 a.a. - 60 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00003214-Q01.10ug
Additional Details : Weight : 0.02000kg
Description
This gene is a member of the Antp homeobox family and encodes a nuclear protein with a homeobox DNA-binding domain. It is included in a cluster of homeobox B genes located on chromosome 17. The encoded protein functions as a sequence-specific transcription factor that is involved in development. Intracellular or ectopic expression of this protein expands hematopoietic stem and progenitor cells in vivo and in vitro, making it a potential candidate for therapeutic stem cell expansion. [provided by RefSeq]
Sequence: MAMSSFLINSNYVDPKFPPCEEYSQSDYLPSDHSPGYYAGGQRRESSFQPEAGFGRRAACSpecifications
NP_076920 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
32.34kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAMSSFLINSNYVDPKFPPCEEYSQSDYLPSDHSPGYYAGGQRRESSFQPEAGFGRRAAC | |
RUO | |
HOXB4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
3214 | |
HOXB4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
HOX-2.6/HOX2/HOX2F | |
HOXB4 | |
Recombinant | |
wheat germ expression system |