missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPR115 Antibody, R&D Systems™

Description
GPR115 Polyclonal specifically detects GPR115 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | GPR115 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | Q8IZF3 |
| Gene Alias | FLJ38076, G protein-coupled receptor 115, G-protein coupled receptor PGR18, PGR18, PGR18G-protein coupled receptor 115, probable G-protein coupled receptor 115, seven transmembrane helix receptor |
| Gene Symbols | GPR115 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?