missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GPER/GPR30 Antibody, R&D Systems™
Mouse Monoclonal Antibody
Brand: R&D Systems MAB5534
This item is not returnable.
View return policy
Description
GPER/GPR30 Monoclonal specifically detects GPER/GPR30 in Human samples. It is validated for Immunohistochemistry.
Specifications
| GPER/GPR30 | |
| Monoclonal | |
| LYOPH | |
| Immunohistochemistry 8-25 ug/mL | |
| Q99527 | |
| GPER | |
| E. coli-derived recombinant human GPER Met1-Ser62 Accession # Q99527 | |
| 100 μg | |
| Primary | |
| Detects human GPER in direct ELISAs. | |
| Human | |
| Purified |
| Immunohistochemistry | |
| 730737 | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CEPR, CEPRG-protein coupled receptor 30, Chemoattractant receptor-like 2, chemokine receptor-like 2, CMKRL2, CMKRL2MGC99678, constitutively expressed peptide-like receptor, DRY12, DRY12IL8-related receptor DRY12, FEG-1, FEG-1mER, Flow-induced endothelial G-protein coupled receptor 1, G protein-coupled estrogen receptor 1, G protein-coupled receptor 30, GPCR-BR, GPR30, GPR30GPCR-Br, G-protein coupled estrogen receptor 1, heptahelix receptor, LERGU, LERGU2, leucine rich protein in GPR30 3'UTR, LyGPR, Lymphocyte-derived G-protein coupled receptor, Membrane estrogen receptor | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2852 | |
| Sterile PBS to a final concentration of 0.5 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction