missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human GILT/IFI30 Antibody, R&D Systems™

Sheep Polyclonal Antibody
Brand: R&D Systems AF7715-SP
This item is not returnable.
View return policy
Description
GILT/IFI30 Polyclonal specifically detects GILT/IFI30 in Human samples. It is validated for Western Blot, Immunohistochemistry.
Specifications
| GILT/IFI30 | |
| Polyclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| EC 1.8, Gamma-interferon-inducible protein IP-30, GILTgamma-interferon-inducible lysosomal thiol reductase, IFI30, IFI-30, interferon, gamma-inducible protein 30, IP30, IP30interferon gamma-inducible protein 30 preproprotein, Legumaturain, MGC32056 | |
| Sheep | |
| Affinity Purified | |
| RUO | |
| 10437 | |
| Sterile PBS to a final concentration of 0.2 mg/mL. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. |
| Western Blot, Immunohistochemistry | |
| LYOPH | |
| Western Blot 0.5 ug/mL, Immunohistochemistry 1-15 ug/mL | |
| P13284 | |
| IFI30 | |
| Human embryonic kidney cell line HEK293-derived recombinant human GILT/IFI30 Ser27-Lys250 Accession # P13284 | |
| 25 μg | |
| Primary | |
| Detects human GILT/IFI30 in direct ELISAs and Western blots. | |
| Human | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction