missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GGA3 Full-length ORF (AAH70044.1, 1 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00023163-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16156992

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Alternative splicing of this gene results in two transcript variants. [provided by RefSeq]

Sequence: MAEAEGESLESWLNKATNPSNRQEDWEYIIGFCDQINKELEGPQIAVRLLAHKIQSPQEWEALQALTYLGDRVSEKVKTKVIELLYSWTMALPEEAKIKDAYHMLKRQGIVQSDPPIPVDRTLIPSPPPRPKNPVFDDEEKSKLLAKLLKSKNPDDLQEANKLIKSMVKEDEARIQKVTKRLHTLEEVNNNVRLLSEMLLHYSQEDSSDGDRELMKELFDQCENKRRTLFKLASETEDNDNSLGDILQASDNLSRVINSYKTIIEGQVINGEVATLTLPDSEGNSQCSNQGTLIDLAELDTTNSLSSVLAPAPTPPSSGIPILPPPPQASGPPRSRSSSQAEATLGPSSTSNALSWLDEELLCLGLADPAPNVPPKESAGNSQWHLLQREQSDLDFFSPRPGTAACGASDAPLLKPSAPSSSSSQAPLPPPFPAPVVPASVPAPSAGSSLFSTGVAPALAPKVEPAVPGHHGLALGNSALHHLDALDQLLEEAKVTSGLVKPTTSPLIPTTTPARPLLPFSTGPGSPLFQPLSFQSQGSPPKGPELSLASIHVPLESIKPSSALPVTAYDKNGFRILFHFAKECPPGRPDVLVVVVSMLNTAPLPVKSIVLQAAVPKSMKVKLQPPSGTELSPFSPIQPPAAITQVMLLANPLKEKVRLRYKLTFALGEQLSTEVGEVDQFPPVEQWGNL
Specifications

Specifications

AAH70044.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
100.9kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
KIAA0154
GGA3
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
23163
GGA3 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAEAEGESLESWLNKATNPSNRQEDWEYIIGFCDQINKELEGPQIAVRLLAHKIQSPQEWEALQALTYLGDRVSEKVKTKVIELLYSWTMALPEEAKIKDAYHMLKRQGIVQSDPPIPVDRTLIPSPPPRPKNPVFDDEEKSKLLAKLLKSKNPDDLQEANKLIKSMVKEDEARIQKVTKRLHTLEEVNNNVRLLSEMLLHYSQEDSSDGDRELMKELFDQCENKRRTLFKLASETEDNDNSLGDILQASDNLSRVINSYKTIIEGQVINGEVATLTLPDSEGNSQCSNQGTLIDLAELDTTNSLSSVLAPAPTPPSSGIPILPPPPQASGPPRSRSSSQAEATLGPSSTSNALSWLDEELLCLGLADPAPNVPPKESAGNSQWHLLQREQSDLDFFSPRPGTAACGASDAPLLKPSAPSSSSSQAPLPPPFPAPVVPASVPAPSAGSSLFSTGVAPALAPKVEPAVPGHHGLALGNSALHHLDALDQLLEEAKVTSGLVKPTTSPLIPTTTPARPLLPFSTGPGSPLFQPLSFQSQGSPPKGPELSLASIHVPLESIKPSSALPVTAYDKNGFRILFHFAKECPPGRPDVLVVVVSMLNTAPLPVKSIVLQAAVPKSMKVKLQPPSGTELSPFSPIQPPAAITQVMLLANPLKEKVRLRYKLTFALGEQLSTEVGEVDQFPPVEQWGNL
RUO
GGA3
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers