Learn More
Abnova™ Human GGA3 Full-length ORF (AAH70044.1, 1 a.a. - 690 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH70044.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 23163 |
Molecular Weight (g/mol) | 100.9kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16166992
|
Abnova™
H00023163-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16156992
|
Abnova™
H00023163-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the Golgi-localized, gamma adaptin ear-containing, ARF-binding (GGA) family. This family includes ubiquitous coat proteins that regulate the trafficking of proteins between the trans-Golgi network and the lysosome. These proteins share an amino-terminal VHS domain which mediates sorting of the mannose 6-phosphate receptors at the trans-Golgi network. They also contain a carboxy-terminal region with homology to the ear domain of gamma-adaptins. Alternative splicing of this gene results in two transcript variants. [provided by RefSeq]
Sequence: MAEAEGESLESWLNKATNPSNRQEDWEYIIGFCDQINKELEGPQIAVRLLAHKIQSPQEWEALQALTYLGDRVSEKVKTKVIELLYSWTMALPEEAKIKDAYHMLKRQGIVQSDPPIPVDRTLIPSPPPRPKNPVFDDEEKSKLLAKLLKSKNPDDLQEANKLIKSMVKEDEARIQKVTKRLHTLEEVNNNVRLLSEMLLHYSQEDSSDGDRELMKELFDQCENKRRTLFKLASETEDNDNSLGDILQASDNLSRVINSYKTIIEGQVINGEVATLTLPDSEGNSQCSNQGTLIDLAELDTTNSLSSVLAPAPTPPSSGIPILPPPPQASGPPRSRSSSQAEATLGPSSTSNALSWLDEELLCLGLADPAPNVPPKESAGNSQWHLLQREQSDLDFFSPRPGTAACGASDAPLLKPSAPSSSSSQAPLPPPFPAPVVPASVPAPSAGSSLFSTGVAPALAPKVEPAVPGHHGLALGNSALHHLDALDQLLEEAKVTSGLVKPTTSPLIPTTTPARPLLPFSTGPGSPLFQPLSFQSQGSPPKGPELSLASIHVPLESIKPSSALPVTAYDKNGFRILFHFAKECPPGRPDVLVVVVSMLNTAPLPVKSIVLQAAVPKSMKVKLQPPSGTELSPFSPIQPPAAITQVMLLANPLKEKVRLRYKLTFALGEQLSTEVGEVDQFPPVEQWGNLSpecifications
AAH70044.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
100.9kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAEAEGESLESWLNKATNPSNRQEDWEYIIGFCDQINKELEGPQIAVRLLAHKIQSPQEWEALQALTYLGDRVSEKVKTKVIELLYSWTMALPEEAKIKDAYHMLKRQGIVQSDPPIPVDRTLIPSPPPRPKNPVFDDEEKSKLLAKLLKSKNPDDLQEANKLIKSMVKEDEARIQKVTKRLHTLEEVNNNVRLLSEMLLHYSQEDSSDGDRELMKELFDQCENKRRTLFKLASETEDNDNSLGDILQASDNLSRVINSYKTIIEGQVINGEVATLTLPDSEGNSQCSNQGTLIDLAELDTTNSLSSVLAPAPTPPSSGIPILPPPPQASGPPRSRSSSQAEATLGPSSTSNALSWLDEELLCLGLADPAPNVPPKESAGNSQWHLLQREQSDLDFFSPRPGTAACGASDAPLLKPSAPSSSSSQAPLPPPFPAPVVPASVPAPSAGSSLFSTGVAPALAPKVEPAVPGHHGLALGNSALHHLDALDQLLEEAKVTSGLVKPTTSPLIPTTTPARPLLPFSTGPGSPLFQPLSFQSQGSPPKGPELSLASIHVPLESIKPSSALPVTAYDKNGFRILFHFAKECPPGRPDVLVVVVSMLNTAPLPVKSIVLQAAVPKSMKVKLQPPSGTELSPFSPIQPPAAITQVMLLANPLKEKVRLRYKLTFALGEQLSTEVGEVDQFPPVEQWGNL | |
RUO | |
GGA3 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
23163 | |
GGA3 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
KIAA0154 | |
GGA3 | |
Recombinant | |
wheat germ expression system |