Learn More
Human GDNF Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 2 publications
Brand:  R&D Systems MAB212-500
 View more versions of this product
Product Code. 18704047
  • 2100.00€ / 500µg
Description
GDNF Monoclonal specifically detects GDNF in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
TRUSTED_SUSTAINABILITY
Specifications
Specifications
GDNF
Monoclonal
LYOPH
Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL
P39905
GDNF
E. coli-derived recombinant human GDNF Arg109-Ile211 Accession # P39905
500 μg
Primary
Detects human GDNF in ELISAs and Western blots. In sandwich ELISAs, no cross-reactivity or interference was observed with recombinant human (rh) BDNF, rhCNTF, rhNeurturin, rh beta-NGF, rhNT-3, or rhNT-4.
Human
Supernatant
Western Blot, ELISA
27106
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
Astrocyte-derived trophic factor, ATF, ATF1, ATF2, glial cell derived neurotrophic factor, glial cell line derived neurotrophic factor, glial cell line-derived neurotrophic factor, HFB1-GDNF, hGDNF, HSCR3
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2668
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.