Learn More
Abnova™ Human GAL3ST4 Partial ORF (NP_078913.3, 388 a.a. - 486 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00079690-Q01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the galactose-3-O-sulfotransferase protein family. The product of this gene catalyzes sulfonation by transferring a sulfate to the C-3' position of galactose residues in O-linked glycoproteins. This enzyme is highly specific for core 1 structures, with asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc being good substrates. [provided by RefSeq]
Sequence: RLQTAVAELRARREALAKHCLVGGEASDPKYITDRRFRPFQFGSAKVLGYILRSGLSPQDQEECERLATPELQYKDKLDAKQFPPTVSLPLKTSRPLSPSpecifications
NP_078913.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
RLQTAVAELRARREALAKHCLVGGEASDPKYITDRRFRPFQFGSAKVLGYILRSGLSPQDQEECERLATPELQYKDKLDAKQFPPTVSLPLKTSRPLSP | |
RUO | |
GAL3ST4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
79690 | |
GAL3ST4 (Human) Recombinant Protein (Q01) | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12116/GAL3ST-4 | |
GAL3ST4 | |
Recombinant | |
wheat germ expression system |