missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human GAL3ST4 Partial ORF (NP_078913.3, 388 a.a. - 486 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_078913.3 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79690 |
Molecular Weight (g/mol) | 36.63kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16100047
|
Abnova™
H00079690-Q01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16190037
|
Abnova™
H00079690-Q01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the galactose-3-O-sulfotransferase protein family. The product of this gene catalyzes sulfonation by transferring a sulfate to the C-3' position of galactose residues in O-linked glycoproteins. This enzyme is highly specific for core 1 structures, with asialofetuin, Gal-beta-1,3-GalNAc and Gal-beta-1,3 (GlcNAc-beta-1,6)GalNAc being good substrates. [provided by RefSeq]
Sequence: RLQTAVAELRARREALAKHCLVGGEASDPKYITDRRFRPFQFGSAKVLGYILRSGLSPQDQEECERLATPELQYKDKLDAKQFPPTVSLPLKTSRPLSPSpecifications
NP_078913.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.63kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ12116/GAL3ST-4 | |
GAL3ST4 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
79690 | |
GAL3ST4 (Human) Recombinant Protein (Q01) | |
RLQTAVAELRARREALAKHCLVGGEASDPKYITDRRFRPFQFGSAKVLGYILRSGLSPQDQEECERLATPELQYKDKLDAKQFPPTVSLPLKTSRPLSP | |
RUO | |
GAL3ST4 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |