missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GADD45A Full-length ORF (AAH11757.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001647-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107017

  • 508.00€ / 25µg
Estimated Shipment: 11-06-2024
to see stock.
Add to basket

Description

Description

This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The DNA damage-induced transcription of this gene is mediated by both p53-dependent and -independent mechanisms. [provided by RefSeq]

Sequence: MTLEEFSAGEQKTERMDKVGDALEEVLSKALSQRTITVGVYEAAKLLNVDPDNVVLCLLAADEDDDRDVALQIHFTLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER
Specifications

Specifications

AAH11757.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DDIT1/GADD45
GADD45A
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1647
GADD45A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MTLEEFSAGEQKTERMDKVGDALEEVLSKALSQRTITVGVYEAAKLLNVDPDNVVLCLLAADEDDDRDVALQIHFTLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER
RUO
GADD45A
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers