missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human GADD45A Full-length ORF (AAH11757.1, 1 a.a. - 165 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

349.00€ - 528.00€

Specifications

Accession Number AAH11757.1
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 1647
Molecular Weight (g/mol) 44.7kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16197007
View Documents Promotion Details
Abnova™
H00001647-P01.10UG
10 ug
349.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
16107017
View Documents Promotion Details
Abnova™
H00001647-P01.25UG
25 ug
528.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 03-11-2023
to see stock.
Add to basket
 
Description

Description

This gene is a member of a group of genes whose transcript levels are increased following stressful growth arrest conditions and treatment with DNA-damaging agents. The protein encoded by this gene responds to environmental stresses by mediating activation of the p38/JNK pathway via MTK1/MEKK4 kinase. The DNA damage-induced transcription of this gene is mediated by both p53-dependent and -independent mechanisms. [provided by RefSeq]

Sequence: MTLEEFSAGEQKTERMDKVGDALEEVLSKALSQRTITVGVYEAAKLLNVDPDNVVLCLLAADEDDDRDVALQIHFTLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER
Specifications

Specifications

AAH11757.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
44.7kDa
Glutathione Sepharose 4 Fast Flow
MTLEEFSAGEQKTERMDKVGDALEEVLSKALSQRTITVGVYEAAKLLNVDPDNVVLCLLAADEDDDRDVALQIHFTLIQAFCCENDINILRVSNPGRLAELLLLETDAGPAASEGAEQPPDLHCVLVTNPHSSQWKDPALSQLICFCRESRYMDQWVPVINLPER
RUO
GADD45A
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
1647
GADD45A (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DDIT1/GADD45
GADD45A
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications