Learn More
Human G-CSFR/CD114 Antibody, R&D Systems™
GREENER_CHOICE
Goat Polyclonal Antibody has been used in 2 publications
Brand:  R&D Systems AF-381-PB
Product Code. 18019161
  • 478.00€ / 100µg
Description
G-CSFR/CD114 Polyclonal specifically detects G-CSFR/CD114 in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
G-CSFR/CD114
Polyclonal
Unconjugated
Q99062
CSF3R
Mouse myeloma cell line NS0-derived recombinant human G-CSF R/CD114 Glu25-Pro621 Accession # Q99062
100 μg
Primary
Detects human G-CSF R/CD114 in direct ELISAs and Western blots. In direct ELISAs, less than 1% cross-reactivity with recombinant human (rh) G-CSF and rhGM-CSF R alpha is observed.
Human
IgG
Western Blot
LYOPH
Western Blot 0.5 ug/mL
CD114, CD114 antigen, colony stimulating factor 3 receptor (granulocyte), CSF3R, Csfgr, G-CSF R, G-CSF receptor, GCSFR, GCSFRG-CSF-R, granulocyte colony-stimulating factor receptor
Goat
Affinity Purified
RUO
1441
Reconstitute at 0.2 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.