Learn More
Abnova™ Human FTSJ2 Partial ORF (NP_037525, 1 a.a. - 103 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00029960-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and it may be involved in the processing and modification of rRNA. This gene has been suggested to be involved in cell cycle control and DNA repair. [provided by RefSeq]
Sequence: MAGYLKLVCVSFQRQGFHTVGSRCKNRTGAEHLWLTRHLRDPFVKAAKVESYRCRSAFKLLEVNERHQILRPGLRVLDCGAAPGAWSQVAVQKVNAAGTDPSSSpecifications
NP_037525 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.07kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MAGYLKLVCVSFQRQGFHTVGSRCKNRTGAEHLWLTRHLRDPFVKAAKVESYRCRSAFKLLEVNERHQILRPGLRVLDCGAAPGAWSQVAVQKVNAAGTDPSS | |
RUO | |
FTSJ2 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
29960 | |
FTSJ2 (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DKFZp686J14194/FJH1 | |
FTSJ2 | |
Recombinant | |
wheat germ expression system |