missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FTSJ2 Partial ORF (NP_037525, 1 a.a. - 103 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number NP_037525
For Use With (Application) Antibody Production, ELISA, Protein Array, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 29960
Molecular Weight (g/mol) 37.07kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16126206
View Documents Promotion Details
Abnova™
H00029960-Q01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16136206
View Documents Promotion Details
Abnova™
H00029960-Q01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

The protein encoded by this gene is a member of the S-adenosylmethionine-binding protein family. It is a nucleolar protein and it may be involved in the processing and modification of rRNA. This gene has been suggested to be involved in cell cycle control and DNA repair. [provided by RefSeq]

Sequence: MAGYLKLVCVSFQRQGFHTVGSRCKNRTGAEHLWLTRHLRDPFVKAAKVESYRCRSAFKLLEVNERHQILRPGLRVLDCGAAPGAWSQVAVQKVNAAGTDPSS
Specifications

Specifications

NP_037525
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.07kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DKFZp686J14194/FJH1
FTSJ2
Recombinant
wheat germ expression system
Antibody Production, ELISA, Protein Array, Western Blot
29960
FTSJ2 (Human) Recombinant Protein (Q01)
MAGYLKLVCVSFQRQGFHTVGSRCKNRTGAEHLWLTRHLRDPFVKAAKVESYRCRSAFKLLEVNERHQILRPGLRVLDCGAAPGAWSQVAVQKVNAAGTDPSS
RUO
FTSJ2
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Product Certifications