Learn More
Abnova™ Human FOXF1 Partial ORF (NP_001442.1, 251 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002294-Q03.25ug
Additional Details : Weight : 0.02000kg
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq]
Sequence: AALNSGASYIKQQPLSPCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDRKEFVFSFNAMASSSMHSAGGGSYYHQQVTYQDIKPCVSpecifications
NP_001442.1 | |
Liquid | |
2294 | |
FOXF1 (Human) Recombinant Protein (Q03) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHL5/FREAC1/MGC105125 | |
FOXF1 | |
Yes | |
wheat germ expression system |
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein) | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
AALNSGASYIKQQPLSPCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDRKEFVFSFNAMASSSMHSAGGGSYYHQQVTYQDIKPCV | |
RUO | |
FOXF1 | |
Wheat Germ (in vitro) | |
GST |