missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Abnova™ Human FOXF1 Partial ORF (NP_001442.1, 251 a.a. - 353 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_001442.1 |
---|---|
For Use With (Application) | Antibody Production, ELISA, Protein Array, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 2294 |
Molecular Weight (g/mol) | 36.96kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16093995
|
Abnova™
H00002294-Q03.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16083995
|
Abnova™
H00002294-Q03.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene belongs to the forkhead family of transcription factors which is characterized by a distinct forkhead domain. The specific function of this gene has not yet been determined; however, it may play a role in the regulation of pulmonary genes as well as embryonic development. [provided by RefSeq]
Sequence: AALNSGASYIKQQPLSPCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDRKEFVFSFNAMASSSMHSAGGGSYYHQQVTYQDIKPCVSpecifications
NP_001442.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
36.96kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FKHL5/FREAC1/MGC105125 | |
FOXF1 | |
Recombinant | |
wheat germ expression system |
Antibody Production, ELISA, Protein Array, Western Blot | |
2294 | |
FOXF1 (Human) Recombinant Protein (Q03) | |
AALNSGASYIKQQPLSPCNPAANPLSGSLSTHSLEQPYLHQNSHNAPAELQGIPRYHSQSPSMCDRKEFVFSFNAMASSSMHSAGGGSYYHQQVTYQDIKPCV | |
RUO | |
FOXF1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |