missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FBXO30 Full-length ORF (NP_115521.3, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084085-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16125623

  • 508.00€ / 25µg
Estimated Shipment: 31-05-2024
to see stock.
Add to basket

Description

Description

This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it is upregulated in nasopharyngeal carcinoma. [provided by RefSeq]

Sequence: MEEELQHSHCVNCVSRRCMTRPEPGISCDLIGCPLVCGAVFHSCKADEHRLLCPFERVPCLNSDFGCPFTMARNKVAEHLEMCPASVVCCTMEWNRWPVSYADRKSYENLSRDVDEVAQLDMALALQDQRMLLESLKVATMMSKATDKVSKPREQISVKSSVPEIPHANGLVSVDEESYGALYQATVETTRSLAAALDILNTATRDIGMLNTSVPNDMDEQQNARESLEDQNLKDQDHLYEEEIGAVGGIDYNDTNQNAQSEQNGSSDLLCDLNTSSYDTSALCNGFPLENICTQVIDQNQNLHGDSKQSNLTNGDCVASSDGTSKPSSSLAVAAQLREIIPSSALPNGTVQHILMPDDEGEGELCWKKVDLGDVKNVDVLSFSHAPSFNFLSNSCWSKPKEDKAVDTSDLEVAEDPMGLQGIDLITAALLFCLGDSPGGRGISDSRMADIYHIDVGTQTFSLPSAILATSTMVGEIASASACDHANPQLSNPSPFQTLGLDLVLECVARYQPKQRSMFTFVCGQLFRRKEFSSHFKNVHGDIHAGLNGWMEQRCPLAYYGCTYSQRRFCPSIQGAKIIHDRHLRSFGVQPCVSTVLVEPARNCVLGLHNDHLSSLPFEVLQHIAGFLDGFSLCQLSCVSKLMRDVCGSLLQSRGMVILQWGKRKYPEGNSSWQIKEKVWRFSTAFCSVNEWKFADILSMADHLKKCSYNVVEKREEAIPLPCMCVTRELTKEGRSLRSVLKPVL
Specifications

Specifications

NP_115521.3
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
108.7kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FLJ41030/Fbx30/MGC21674
FBXO30
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84085
FBXO30 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MEEELQHSHCVNCVSRRCMTRPEPGISCDLIGCPLVCGAVFHSCKADEHRLLCPFERVPCLNSDFGCPFTMARNKVAEHLEMCPASVVCCTMEWNRWPVSYADRKSYENLSRDVDEVAQLDMALALQDQRMLLESLKVATMMSKATDKVSKPREQISVKSSVPEIPHANGLVSVDEESYGALYQATVETTRSLAAALDILNTATRDIGMLNTSVPNDMDEQQNARESLEDQNLKDQDHLYEEEIGAVGGIDYNDTNQNAQSEQNGSSDLLCDLNTSSYDTSALCNGFPLENICTQVIDQNQNLHGDSKQSNLTNGDCVASSDGTSKPSSSLAVAAQLREIIPSSALPNGTVQHILMPDDEGEGELCWKKVDLGDVKNVDVLSFSHAPSFNFLSNSCWSKPKEDKAVDTSDLEVAEDPMGLQGIDLITAALLFCLGDSPGGRGISDSRMADIYHIDVGTQTFSLPSAILATSTMVGEIASASACDHANPQLSNPSPFQTLGLDLVLECVARYQPKQRSMFTFVCGQLFRRKEFSSHFKNVHGDIHAGLNGWMEQRCPLAYYGCTYSQRRFCPSIQGAKIIHDRHLRSFGVQPCVSTVLVEPARNCVLGLHNDHLSSLPFEVLQHIAGFLDGFSLCQLSCVSKLMRDVCGSLLQSRGMVILQWGKRKYPEGNSSWQIKEKVWRFSTAFCSVNEWKFADILSMADHLKKCSYNVVEKREEAIPLPCMCVTRELTKEGRSLRSVLKPVL
RUO
FBXO30
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers