Learn More
Abnova™ Human FBXO30 Full-length ORF (NP_115521.3, 1 a.a. - 745 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | NP_115521.3 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84085 |
Molecular Weight (g/mol) | 108.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16115623
|
Abnova™
H00084085-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
16125623
|
Abnova™
H00084085-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 10-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the F-box protein family which is characterized by an approximately 40 amino acid motif, the F-box. The F-box proteins constitute one of the four subunits of the ubiquitin protein ligase complex called SCFs (SKP1-cullin-F-box), which function in phosphorylation-dependent ubiquitination. The F-box proteins are divided into 3 classes: Fbws containing WD-40 domains, Fbls containing leucine-rich repeats, and Fbxs containing either different protein-protein interaction modules or no recognizable motifs. The protein encoded by this gene belongs to the Fbxs class and it is upregulated in nasopharyngeal carcinoma. [provided by RefSeq]
Sequence: MEEELQHSHCVNCVSRRCMTRPEPGISCDLIGCPLVCGAVFHSCKADEHRLLCPFERVPCLNSDFGCPFTMARNKVAEHLEMCPASVVCCTMEWNRWPVSYADRKSYENLSRDVDEVAQLDMALALQDQRMLLESLKVATMMSKATDKVSKPREQISVKSSVPEIPHANGLVSVDEESYGALYQATVETTRSLAAALDILNTATRDIGMLNTSVPNDMDEQQNARESLEDQNLKDQDHLYEEEIGAVGGIDYNDTNQNAQSEQNGSSDLLCDLNTSSYDTSALCNGFPLENICTQVIDQNQNLHGDSKQSNLTNGDCVASSDGTSKPSSSLAVAAQLREIIPSSALPNGTVQHILMPDDEGEGELCWKKVDLGDVKNVDVLSFSHAPSFNFLSNSCWSKPKEDKAVDTSDLEVAEDPMGLQGIDLITAALLFCLGDSPGGRGISDSRMADIYHIDVGTQTFSLPSAILATSTMVGEIASASACDHANPQLSNPSPFQTLGLDLVLECVARYQPKQRSMFTFVCGQLFRRKEFSSHFKNVHGDIHAGLNGWMEQRCPLAYYGCTYSQRRFCPSIQGAKIIHDRHLRSFGVQPCVSTVLVEPARNCVLGLHNDHLSSLPFEVLQHIAGFLDGFSLCQLSCVSKLMRDVCGSLLQSRGMVILQWGKRKYPEGNSSWQIKEKVWRFSTAFCSVNEWKFADILSMADHLKKCSYNVVEKREEAIPLPCMCVTRELTKEGRSLRSVLKPVLSpecifications
NP_115521.3 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
108.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MEEELQHSHCVNCVSRRCMTRPEPGISCDLIGCPLVCGAVFHSCKADEHRLLCPFERVPCLNSDFGCPFTMARNKVAEHLEMCPASVVCCTMEWNRWPVSYADRKSYENLSRDVDEVAQLDMALALQDQRMLLESLKVATMMSKATDKVSKPREQISVKSSVPEIPHANGLVSVDEESYGALYQATVETTRSLAAALDILNTATRDIGMLNTSVPNDMDEQQNARESLEDQNLKDQDHLYEEEIGAVGGIDYNDTNQNAQSEQNGSSDLLCDLNTSSYDTSALCNGFPLENICTQVIDQNQNLHGDSKQSNLTNGDCVASSDGTSKPSSSLAVAAQLREIIPSSALPNGTVQHILMPDDEGEGELCWKKVDLGDVKNVDVLSFSHAPSFNFLSNSCWSKPKEDKAVDTSDLEVAEDPMGLQGIDLITAALLFCLGDSPGGRGISDSRMADIYHIDVGTQTFSLPSAILATSTMVGEIASASACDHANPQLSNPSPFQTLGLDLVLECVARYQPKQRSMFTFVCGQLFRRKEFSSHFKNVHGDIHAGLNGWMEQRCPLAYYGCTYSQRRFCPSIQGAKIIHDRHLRSFGVQPCVSTVLVEPARNCVLGLHNDHLSSLPFEVLQHIAGFLDGFSLCQLSCVSKLMRDVCGSLLQSRGMVILQWGKRKYPEGNSSWQIKEKVWRFSTAFCSVNEWKFADILSMADHLKKCSYNVVEKREEAIPLPCMCVTRELTKEGRSLRSVLKPVL | |
RUO | |
FBXO30 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84085 | |
FBXO30 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FLJ41030/Fbx30/MGC21674 | |
FBXO30 | |
Recombinant | |
wheat germ expression system |