missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human FA2H Full-length ORF (AAH17049.2, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00079152-P01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114123

  • 508.00€ / 25µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

Sphingolipids are a large class of lipids found in all eukaryotic cells and are involved in numerous cellular processes. The structural diversity of sphingolipids stems from more than 300 distinct head groups, as well as from modifications of the hydrophobic ceramide moiety. FA2H catalyzes a common modification of the ceramide moiety: hydroxylation at the 2 position of the N-acyl chain. Sphingolipids containing 2-hydroxy fatty acid are common in nervous and epidermal tissue. Glycosphingolipids containing a high proportion of 2-hydroxy fatty acid are critical components of myelin, and several very long chain ceramides with 2-hydroxy fatty acids are important for the permeability barrier function of epidermis (Alderson et al., 2004 [PubMed 15337768]).[supplied by OMIM]

Sequence: MAPAPPPAASFSPSEVQRRLAAGACWVRRGARLYDLSSFVRHHPGGEQLLRARAGQDISADLDGPPHRHSANARRWLEQYYVGELRGEQQGSMENEPVALEETQKTDPAMEPRFKVVDWDKDLVDWRKPLLWQVGHLGEKYDEWVHQPVTRPIRLFHSDLIEGLSKTVWYSVPIIWVPLVLYLSWSYYRTFAQGNVRLFTSFTTEYTVAVPKSMFPGLFMLGTFLWSLIEYLIHRFLFHMKPPSDSYYLIMLHFVMHGQHHKAPFDGSRLVFPPVPASLVIGVFYLCMQLILPEAVGGTVFAGGLLGYVLYDMTHYYLHFGSPHKGSYLYSLKAHHVKHHFAHQKSGFGISTKLWDYCFHTLTPEKPHLKTQ
Specifications

Specifications

AAH17049.2
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
69.2kDa
Glutathione Sepharose 4 Fast Flow
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
FAAH/FAH1/FAXDC1/FLJ25287/SCS7
FA2H
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
79152
FA2H (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAPAPPPAASFSPSEVQRRLAAGACWVRRGARLYDLSSFVRHHPGGEQLLRARAGQDISADLDGPPHRHSANARRWLEQYYVGELRGEQQGSMENEPVALEETQKTDPAMEPRFKVVDWDKDLVDWRKPLLWQVGHLGEKYDEWVHQPVTRPIRLFHSDLIEGLSKTVWYSVPIIWVPLVLYLSWSYYRTFAQGNVRLFTSFTTEYTVAVPKSMFPGLFMLGTFLWSLIEYLIHRFLFHMKPPSDSYYLIMLHFVMHGQHHKAPFDGSRLVFPPVPASLVIGVFYLCMQLILPEAVGGTVFAGGLLGYVLYDMTHYYLHFGSPHKGSYLYSLKAHHVKHHFAHQKSGFGISTKLWDYCFHTLTPEKPHLKTQ
RUO
FA2H
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers