Learn More
Abnova™ Human FA2H Full-length ORF (AAH17049.2, 1 a.a. - 372 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH17049.2 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 79152 |
Molecular Weight (g/mol) | 69.2kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16114123
|
Abnova™
H00079152-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16104123
|
Abnova™
H00079152-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
Sphingolipids are a large class of lipids found in all eukaryotic cells and are involved in numerous cellular processes. The structural diversity of sphingolipids stems from more than 300 distinct head groups, as well as from modifications of the hydrophobic ceramide moiety. FA2H catalyzes a common modification of the ceramide moiety: hydroxylation at the 2 position of the N-acyl chain. Sphingolipids containing 2-hydroxy fatty acid are common in nervous and epidermal tissue. Glycosphingolipids containing a high proportion of 2-hydroxy fatty acid are critical components of myelin, and several very long chain ceramides with 2-hydroxy fatty acids are important for the permeability barrier function of epidermis (Alderson et al., 2004 [PubMed 15337768]).[supplied by OMIM]
Sequence: MAPAPPPAASFSPSEVQRRLAAGACWVRRGARLYDLSSFVRHHPGGEQLLRARAGQDISADLDGPPHRHSANARRWLEQYYVGELRGEQQGSMENEPVALEETQKTDPAMEPRFKVVDWDKDLVDWRKPLLWQVGHLGEKYDEWVHQPVTRPIRLFHSDLIEGLSKTVWYSVPIIWVPLVLYLSWSYYRTFAQGNVRLFTSFTTEYTVAVPKSMFPGLFMLGTFLWSLIEYLIHRFLFHMKPPSDSYYLIMLHFVMHGQHHKAPFDGSRLVFPPVPASLVIGVFYLCMQLILPEAVGGTVFAGGLLGYVLYDMTHYYLHFGSPHKGSYLYSLKAHHVKHHFAHQKSGFGISTKLWDYCFHTLTPEKPHLKTQSpecifications
AAH17049.2 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
69.2kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAPAPPPAASFSPSEVQRRLAAGACWVRRGARLYDLSSFVRHHPGGEQLLRARAGQDISADLDGPPHRHSANARRWLEQYYVGELRGEQQGSMENEPVALEETQKTDPAMEPRFKVVDWDKDLVDWRKPLLWQVGHLGEKYDEWVHQPVTRPIRLFHSDLIEGLSKTVWYSVPIIWVPLVLYLSWSYYRTFAQGNVRLFTSFTTEYTVAVPKSMFPGLFMLGTFLWSLIEYLIHRFLFHMKPPSDSYYLIMLHFVMHGQHHKAPFDGSRLVFPPVPASLVIGVFYLCMQLILPEAVGGTVFAGGLLGYVLYDMTHYYLHFGSPHKGSYLYSLKAHHVKHHFAHQKSGFGISTKLWDYCFHTLTPEKPHLKTQ | |
RUO | |
FA2H | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
79152 | |
FA2H (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
FAAH/FAH1/FAXDC1/FLJ25287/SCS7 | |
FA2H | |
Recombinant | |
wheat germ expression system |