missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human EZFIT aa 6-199 Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems PP-K9716-00
This item is not returnable.
View return policy
Description
EZFIT/ZNF71 Monoclonal specifically detects EZFIT/ZNF71 in Human samples. It is validated for Western Blot, Direct ELISA.
Specifications
| EZFIT/ZNF71 | |
| Monoclonal | |
| Unconjugated | |
| A liquid formulation in physiologic saline with 0.1% NaN 3 . with 0.1% Sodium Azide | |
| ZNF71 | |
| aa 6-199 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Western Blot, ELISA | |
| K9716 | |
| Western Blot, Direct ELISA | |
| Cos26, endothelial zinc finger protein induced by tumor necrosis factor alpha, Kruppel-related zinc finger protein, zinc finger protein 71 (Cos26), zinc finger protein 71EZFITCos26, ZNF47, ZNF71 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 58491 | |
| Store the unopened product at -70 degreesC. Use a manual defrost freezer and avoid repeated freeze-thaw cycles. Do not use past expiration date. | |
| IgG2a |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction