missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 5 publications
Brand: R&D Systems MAB287-100
This item is not returnable.
View return policy
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Neutralization.
Specifications
| Erythropoietin/EPO | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2 | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 2056 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG1 |
| Neutralization | |
| 9C21D11 | |
| Neutralization 0.5-1.5 ug/mL | |
| P01588 | |
| EPO | |
| Chinese hamster ovary cell line CHO-derived recombinant human Erythropoietin/EPO, Accession # P01588 | |
| 100 μg | |
| Primary | |
| Detects human Erythropoietin/EPO. | |
| Human | |
| Supernatant |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction