Learn More
Human Erythropoietin/EPO Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody has been used in 5 publications
Brand:  R&D Systems MAB287-SP
 View more versions of this product
Product Code. 18749427
  • ONLINE EXCLUSIVE PRICE
    209.00€
    197.40€ / 25µg
    Save 11.60€
    5% Off
Description
Erythropoietin/EPO Monoclonal specifically detects Erythropoietin/EPO in Human samples. It is validated for Neutralization.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Erythropoietin/EPO
Monoclonal
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
ECYT5, EP, EPO, epoetin, erythropoietin, MGC138142, MVCD2
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
2056
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG1
Neutralization
9C21D11
Neutralization 0.5-1.5 ug/mL
P01588
EPO
Chinese hamster ovary cell line CHO-derived recombinant human Erythropoietin/EPO, Accession # P01588
25 μg
Primary
Detects human Erythropoietin/EPO.