Learn More
Abnova™ Human ERF Partial ORF (AAH22231, 181 a.a. - 290 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00002077-Q01.25ug
Additional Details : Weight : 0.00010kg
Description
Members of the ETS family of transcription factors, such as ERF, regulate cell proliferation and differentiation. They share a highly conserved DNA-binding domain, the ETS domain, that recognizes the sequence GGAA/T (de Castro et al., 1997 [PubMed 9192842]). For further information on ETS transcription factors, see ETS1 (MIM 164720).[supplied by OMIM]
Sequence: LGRGSVSDCSDGTSELEEPLGEDPRARPPGPPDLGAFRGPPLARLPHDPGVFRVYPRPRGGPEPLSPFPVSPLAGPGSLLPPQLSPALPMTPTHLAYTPSPTLSPMYPSGSpecifications
AAH22231 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
37.73kDa | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
LGRGSVSDCSDGTSELEEPLGEDPRARPPGPPDLGAFRGPPLARLPHDPGVFRVYPRPRGGPEPLSPFPVSPLAGPGSLLPPQLSPALPMTPTHLAYTPSPTLSPMYPSG | |
RUO | |
ERF | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, ELISA, Protein Array, Western Blot | |
2077 | |
ERF (Human) Recombinant Protein (Q01) | |
25 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
PE-2/PE2 | |
ERF | |
Recombinant | |
wheat germ expression system |