missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human ErbB4/Her4 Alexa Fluor™ 750-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody
Brand: R&D Systems FAB11311S-100UG
This item is not returnable.
View return policy
Description
ErbB4/Her4 Monoclonal specifically detects ErbB4/Her4 in Human samples. It is validated for Flow Cytometry.
Specifications
| ErbB4/Her4 | |
| Monoclonal | |
| 0.2 mg/mL | |
| Flow Cytometry 0.25-1 ug/10^6 cells | |
| Q15303 | |
| ERBB4 | |
| Mouse myeloma cell line NS0-derived recombinant human ErbB4/Her4 Glu26-Arg649 Accession # Q15303 | |
| RUO | |
| 2066 | |
| Human | |
| IgG2a |
| Flow Cytometry | |
| 182818 | |
| Alexa Fluor 750 | |
| Supplied 0.2 mg/mL in a saline solution containing BSA and Sodium Azide. with 0.09% Sodium Azide | |
| avian erythroblastic leukemia viral (v-erb-b2) oncogene homolog 4, EC 2.7.10, EC 2.7.10.1, HER4, HER4MGC138404, p180erbB4, Proto-oncogene-like protein c-ErbB-4, receptor tyrosine-protein kinase erbB-4, Tyrosine kinase-type cell surface receptor HER4, v-erb-a avian erythroblastic leukemia viral oncogene homolog-like 4, v-erb-a erythroblastic leukemia viral oncogene homolog 4 (avian) | |
| Mouse | |
| 100 μg | |
| Primary | |
| Detects human ErbB4/Her4 in direct ELISAs and Western blots. In direct ELISAs and Western blots, no cross-reactivity with recombinant human (rh) EGF R, rhErbB2, or rhErbB3 is observed. | |
| Store the unopened product at 2 - 8 degreesC. Do not use past expiration date. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction