missing translation for 'onlineSavingsMsg'
Learn More

Human ErbB2/Her2 (Research Grade Trastuzumab Biosimilar) Antibody, R&D Systems™

Product Code. 18713859
Change view
Click to view available options
Quantity:
100 μg
25 μg
Unit Size:
100µg
25µg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
18713859 25 μg 25µg
18769767 100 μg 100µg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price
Product Code. 18713859 Supplier R&D Systems Supplier No. MAB9589SP
  • ONLINE EXCLUSIVE PRICE
    227.00€
    214.20€ / 25µg
    Save 12.80€
    5% Off
View all promotions
 Restricted Item
Estimated Shipment: 02-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy
To receive the discount customers must purchase three of the same product at list price in a single order to receive 33.33% discount. There is no limit to the multiples of 3 that customers can buy. Use promo code ”27950” to get your promotional price

Human Monoclonal Antibody

ErbB2/Her2 Monoclonal specifically detects ErbB2/Her2 in Human∣Staphylococcus∣Streptococcus samples. It is validated for CyTOF∣ELISA Detection∣Flow Cytometry∣Immunoprecipitation∣Inhibition of Cell Growth∣Neutralization Control∣Western Blot.
TRUSTED_SUSTAINABILITY

Specifications

Antigen ErbB2/Her2
Applications CyTOF, ELISA Detection (Matched Antibody Pair), Flow Cytometry, Immunoprecipitation, Inhibition Assays, Neutralization Control, Western Blot
Classification Monoclonal
Clone Hu5
Conjugate Unconjugated
Dilution Flow Cytometry 0.25 ug/10^6 cells, Inhibition of Cell Growth 15-75 ng/mL, CyTOF-ready
Formulation Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS.
Gene Alias CD340, CD340 antigen, c-erb B2/neu protein, EC 2.7.10, EGFR2, HER2, HER-2, HER2EC 2.7.10.1, herstatin, Metastatic lymph node gene 19 protein, MLN 19, MLN19, Neu Oncogene, NEUHER-2/neu, neuroblastoma/glioblastoma derived oncogene homolog, NGL, NGLTKR1, p185erbB2, Proto-oncogene c-ErbB-2, Proto-oncogene Neu, receptor tyrosine-protein kinase erbB-2, TKR1, Tyrosine kinase-type cell surface receptor HER2, v-erb-b2 avian erythroblastic leukemia viral oncogene homolog 2(neuro/glioblastoma derived oncogene homolog), v-erb-b2 erythroblastic leukemia viral oncogene homolog 2, neuro/glioblastomaderived oncogene homolog (avian)
Host Species Human
Immunogen Human ErbB2/Her2
Purification Method Protein A or G purified from cell culture supernatant
Quantity 25 μg
Primary or Secondary Primary
Gene ID (Entrez) 2064
Test Specificity Detects human ErbB2/Her2 in direct ELISAs. This non-therapeutic antibody uses the same variable region sequence as the therapeutic antibody Trastuzumab. This product is for research use only.
Reconstitution Reconstitute at 0.5 mg/mL in sterile PBS.
Target Species Human, Staphylococcus species, Streptococcus species
Content And Storage Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
Form Supernatant
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.