missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPHA4 Full-length ORF (no Protein_acc, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00002043-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16114581

  • 335.00€ / 10µg
Estimated Shipment: 07-06-2024
to see stock.
Add to basket

Description

Description

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]

Sequence: MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
Specifications

Specifications

no protein_acc
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HEK8/SEK/TYRO1
EPHA4
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
2043
EPHA4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
RUO
EPHA4
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers