missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human EPHA4 Full-length ORF (no Protein_acc, 1 a.a. - 105 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

335.00€ - 508.00€

Specifications

Accession Number no protein_acc
For Use With (Application) Antibody Production, Protein Array, ELISA, Western Blot
Formulation 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
Gene ID (Entrez) 2043
Molecular Weight (g/mol) 37.18kDa
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
16124581
View Documents Promotion Details
Abnova™
H00002043-P01.25UG
25 ug
508.00€
25µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
16114581
View Documents Promotion Details
Abnova™
H00002043-P01.10UG
10 ug
335.00€
10µg
View alternative products
Save Up to 
»
Estimated Shipment: 27-05-2024
to see stock.
Add to basket
 
Description

Description

This gene belongs to the ephrin receptor subfamily of the protein-tyrosine kinase family. EPH and EPH-related receptors have been implicated in mediating developmental events, particularly in the nervous system. Receptors in the EPH subfamily typically have a single kinase domain and an extracellular region containing a Cys-rich domain and 2 fibronectin type III repeats. The ephrin receptors are divided into 2 groups based on the similarity of their extracellular domain sequences and their affinities for binding ephrin-A and ephrin-B ligands. [provided by RefSeq]

Sequence: MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
Specifications

Specifications

no protein_acc
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.18kDa
Glutathione Sepharose 4 Fast Flow
MCVHRCECVCMRACLCAGVCMCIASCLGLPMNVVECYTWRVLVFHQFQDEELHDTVDLETIPLERQPRDVQHPVSTRILYLHVYFVAVTLTLIRILQLWTEAFSP
RUO
EPHA4
Wheat Germ (in vitro)
GST
Liquid
Antibody Production, Protein Array, ELISA, Western Blot
2043
EPHA4 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
HEK8/SEK/TYRO1
EPHA4
Recombinant
wheat germ expression system
Videos
SDS
Documents

Documents

Product Certifications