Learn More
ACROBiosystems Human EGF R Protein, His Tag, low endotoxin
low endotoxin
Brand: ACROBiosystems EGR-H522a-100ug
Additional Details : Weight : 0.00010kg
Description
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5kDa. The protein migrates as 110-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_005219.2 | |
100 μg | |
Leu 25 - Ser 645 | |
EGFR,ERBB,ERBB1,HER1,PIG61,mENA | |
Unconjugated | |
His Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |
70.5 kDa | |
Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). | |
-20°C | |
EGF R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |