missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ACROBiosystems Human EGF R Protein, His Tag, low endotoxin
low endotoxin
660.77€ - 3459.34€
Specifications
Accession Number | NP_005219.2 |
---|---|
Molecular Weight (g/mol) | 70.5 kDa |
Source | Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). |
Immunogen | Leu 25 - Ser 645 |
Storage Requirements | -20°C |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16437489
|
ACROBiosystems
EGR-H522A-100UG |
100 μg |
660.77€
100µg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
15816627
|
ACROBiosystems
EGR-H522A-1MG |
1 mg |
3459.34€
1mg |
Estimated Shipment: 11-04-2024
Log in to see stock. |
|||||
Description
The epidermal growth factor receptor (EGFR; ErbB-1; HER1 in humans) is the cell-surface receptor for members of the epidermal growth factor family (EGF-family) of extracellular protein ligands. The epidermal growth factor receptor is a member of the ErbB family of receptors, a subfamily of four closely related receptor tyrosine kinases: EGFR (ErbB-1), HER2/c-neu (ErbB-2), Her 3 (ErbB-3) and Her 4 (ErbB-4). Mutations affecting EGFR expression or activity could result in cancer.
This protein carries a polyhistidine tag at the C-terminus. The protein has a calculated MW of 70.5kDa. The protein migrates as 110-115 kDa under reducing (R) condition (SDS-PAGE) due to glycosylation.Specifications
NP_005219.2 | |
Human EGF R, His Tag, low endotoxin (EGR-H522a) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Ser 645 (Accession # P00533-1). | |
-20°C | |
EGF R | |
Human | |
PBS, pH7.4 | |
HEK293 | |
Bio-Markers and CD Antigens |
70.5 kDa | |
Leu 25 - Ser 645 | |
EGFR,ERBB,ERBB1,HER1,PIG61,mENA | |
Unconjugated | |
His Tag | |
Less than 0.01 EU / μg by the LAL method. | |
Powder | |
>95% as determined by SDS-PAGE |