missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human E-Selectin/P-Selectin (CD62E/P) Fluorescein-conjugated Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems FAB6169F
This item is not returnable.
View return policy
Description
E-Selectin (CD62E)/P-Selectin (CD62P) Monoclonal specifically detects E-Selectin (CD62E)/P-Selectin (CD62P) in Human samples. It is validated for Flow Cytometry.
Specifications
| E-Selectin (CD62E)/P-Selectin (CD62P) | |
| Monoclonal | |
| Fluorescein | |
| Supplied in a saline solution containing BSA and Sodium Azide. with Sodium Azide | |
| Mouse | |
| Affinity Purified | |
| RUO | |
| 6401 | |
| Human | |
| IgG1 |
| Flow Cytometry | |
| BBIG-E6 (13D5) | |
| Flow Cytometry 10 uL/10^6 cells | |
| CD62E, CD62P, ELAM, ELAM1, ESEL, Eselectin, GMP140, GRMP, LECAM2, PADGEM, PSEL, PSelectin, SELE, SELP | |
| Activated HUVEC human umbilical vein endothelial cells | |
| 100 Tests | |
| Primary | |
| Detects human E-Selectin/P-Selectin (CD62E/P). Binds to COS cells transfected with human E-Selectin or human P-Selectin. It does not bind to CHO cells transfected with human ICAM-1, L-Selectin, PECAM-1 or VCAM-1. | |
| Protect from light. Do not freeze. 12 months from date of receipt, 2 to 8 degreesC as supplied. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction