Learn More
Human Desmocollin-1 Antibody, R&D Systems™
Rabbit Monoclonal Antibody
Brand: R&D Systems MAB4955-100
This item is not returnable.
View return policy
Description
Desmocollin-1 Monoclonal antibody specifically detects Desmocollin-1 in Human samples. It is validated for Immunohistochemistry
Specifications
| Desmocollin-1 | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied either lyophilized or as a 0.2 μm filtered solution in PBS. | |
| cadherin family member 1, CDHF1, desmocollin 1, desmocollin-1, Desmocollin1, desmosomal glycoprotein 2/3, DG2/DG3, DSC1 | |
| Mouse myeloma cell line, NS0-derived human Desmocollin-1, Arg135-Asn686, Accession # Q08554 | |
| 100 μg | |
| Primary | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 °C as supplied. 1 month, 2 to 8 °C under sterile conditions after reconstitution. 6 months, -20 to -70 °C under sterile conditions after reconstitution. | |
| IgG |
| Immunohistochemistry | |
| 2906A | |
| Immunohistochemistry 3-25 ug/mL | |
| Q08554 | |
| Rabbit | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1823 | |
| Human | |
| Purified |
Product Content Correction
Product Title
Haben Sie Verbesserungsvorschläge?Übermitteln Sie eine inhaltliche Korrektur