missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DDX1 Full-length ORF (AAH12132, 1 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001653-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16163981

  • 335.00€ / 10µg
Estimated Shipment: 12-06-2024
to see stock.
Add to basket

Description

Description

DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein of unknown function. It shows high transcription levels in 2 retinoblastoma cell lines and in tissues of neuroectodermal origin. [provided by RefSeq]

Sequence: MAAFSEMGVMPEIAQAVEEMDWLLPTDIQAESIPLILGGGDVLMAAETGSGKTGAFSIPVIQIVYETLKDQQEGKKGKTTIKTGASVLNKWQMNPYDRGSAFAIGSDGLCCQSREVKEWHGCRATKGLMKGKHYYEVSCHDQGLCRVGWSTMQASLDLGTDKFGFGFGGTGKKSHNKQFDNYGEEFTMHDTIGCYLDIDKGHVKFSKNGKDLGLAFEIPPHMKNQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKYIDNPKLRELLIIGGVAARDQLSVLENGVDIVVGTPGRLDDLVSTGKLNLSQVRFLVLDEADGLLSQGYSDFINRMHNQIPQVTSDGKRLQVIVCSATLHSFDVKKLSEKIMHFPTWVDLKGEDSVPDTVHHVVVPVNPKTDRLWERLGKSHIRTDDVHAKDNTRPGANSPEMWSEAIKILKGEYAVRAIKEHKMDQAIIFCRTKIDCDNLEQYFIQQGGGPDKKGHQFSCVCLHGDRKPHERKQNLERFKKGDVRFLICTDVAARGIDIHGVPYVINVTLPDEKQNYVHRIGRVGRAERMGLAISLVATEKEKVWYHVCSSRGKGCYNTRLKEDGGCTIWYNEMQLLSEIEEHLNCTISQVEPDIKVPVDEFDGKVTYGQKRAAGGGSYKGHVDILAPTVQELAALEKEAQTSFLHLGYLPNQLFRTF
Specifications

Specifications

AAH12132
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
107.14kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DBP-RB/UKVH5d
DDX1
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
1653
DDX1 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MAAFSEMGVMPEIAQAVEEMDWLLPTDIQAESIPLILGGGDVLMAAETGSGKTGAFSIPVIQIVYETLKDQQEGKKGKTTIKTGASVLNKWQMNPYDRGSAFAIGSDGLCCQSREVKEWHGCRATKGLMKGKHYYEVSCHDQGLCRVGWSTMQASLDLGTDKFGFGFGGTGKKSHNKQFDNYGEEFTMHDTIGCYLDIDKGHVKFSKNGKDLGLAFEIPPHMKNQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKYIDNPKLRELLIIGGVAARDQLSVLENGVDIVVGTPGRLDDLVSTGKLNLSQVRFLVLDEADGLLSQGYSDFINRMHNQIPQVTSDGKRLQVIVCSATLHSFDVKKLSEKIMHFPTWVDLKGEDSVPDTVHHVVVPVNPKTDRLWERLGKSHIRTDDVHAKDNTRPGANSPEMWSEAIKILKGEYAVRAIKEHKMDQAIIFCRTKIDCDNLEQYFIQQGGGPDKKGHQFSCVCLHGDRKPHERKQNLERFKKGDVRFLICTDVAARGIDIHGVPYVINVTLPDEKQNYVHRIGRVGRAERMGLAISLVATEKEKVWYHVCSSRGKGCYNTRLKEDGGCTIWYNEMQLLSEIEEHLNCTISQVEPDIKVPVDEFDGKVTYGQKRAAGGGSYKGHVDILAPTVQELAALEKEAQTSFLHLGYLPNQLFRTF
RUO
DDX1
Wheat Germ (in vitro)
GST
Liquid
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers