Learn More
Abnova™ Human DDX1 Full-length ORF (AAH12132, 1 a.a. - 740 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH12132 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 1653 |
Molecular Weight (g/mol) | 107.14kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16173981
|
Abnova™
H00001653-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
16163981
|
Abnova™
H00001653-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 31-05-2024
Log in to see stock. |
|||||
Description
DEAD box proteins, characterized by the conserved motif Asp-Glu-Ala-Asp (DEAD), are putative RNA helicases. They are implicated in a number of cellular processes involving alteration of RNA secondary structure such as translation initiation, nuclear and mitochondrial splicing, and ribosome and spliceosome assembly. Based on their distribution patterns, some members of this family are believed to be involved in embryogenesis, spermatogenesis, and cellular growth and division. This gene encodes a DEAD box protein of unknown function. It shows high transcription levels in 2 retinoblastoma cell lines and in tissues of neuroectodermal origin. [provided by RefSeq]
Sequence: MAAFSEMGVMPEIAQAVEEMDWLLPTDIQAESIPLILGGGDVLMAAETGSGKTGAFSIPVIQIVYETLKDQQEGKKGKTTIKTGASVLNKWQMNPYDRGSAFAIGSDGLCCQSREVKEWHGCRATKGLMKGKHYYEVSCHDQGLCRVGWSTMQASLDLGTDKFGFGFGGTGKKSHNKQFDNYGEEFTMHDTIGCYLDIDKGHVKFSKNGKDLGLAFEIPPHMKNQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKYIDNPKLRELLIIGGVAARDQLSVLENGVDIVVGTPGRLDDLVSTGKLNLSQVRFLVLDEADGLLSQGYSDFINRMHNQIPQVTSDGKRLQVIVCSATLHSFDVKKLSEKIMHFPTWVDLKGEDSVPDTVHHVVVPVNPKTDRLWERLGKSHIRTDDVHAKDNTRPGANSPEMWSEAIKILKGEYAVRAIKEHKMDQAIIFCRTKIDCDNLEQYFIQQGGGPDKKGHQFSCVCLHGDRKPHERKQNLERFKKGDVRFLICTDVAARGIDIHGVPYVINVTLPDEKQNYVHRIGRVGRAERMGLAISLVATEKEKVWYHVCSSRGKGCYNTRLKEDGGCTIWYNEMQLLSEIEEHLNCTISQVEPDIKVPVDEFDGKVTYGQKRAAGGGSYKGHVDILAPTVQELAALEKEAQTSFLHLGYLPNQLFRTFSpecifications
AAH12132 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
107.14kDa | |
Glutathione Sepharose 4 Fast Flow | |
MAAFSEMGVMPEIAQAVEEMDWLLPTDIQAESIPLILGGGDVLMAAETGSGKTGAFSIPVIQIVYETLKDQQEGKKGKTTIKTGASVLNKWQMNPYDRGSAFAIGSDGLCCQSREVKEWHGCRATKGLMKGKHYYEVSCHDQGLCRVGWSTMQASLDLGTDKFGFGFGGTGKKSHNKQFDNYGEEFTMHDTIGCYLDIDKGHVKFSKNGKDLGLAFEIPPHMKNQALFPACVLKNAELKFNFGEEEFKFPPKDGFVALSKAPDGYIVKSQHSGNAQVTQTKFLPNAPKALIVEPSRELAEQTLNNIKQFKKYIDNPKLRELLIIGGVAARDQLSVLENGVDIVVGTPGRLDDLVSTGKLNLSQVRFLVLDEADGLLSQGYSDFINRMHNQIPQVTSDGKRLQVIVCSATLHSFDVKKLSEKIMHFPTWVDLKGEDSVPDTVHHVVVPVNPKTDRLWERLGKSHIRTDDVHAKDNTRPGANSPEMWSEAIKILKGEYAVRAIKEHKMDQAIIFCRTKIDCDNLEQYFIQQGGGPDKKGHQFSCVCLHGDRKPHERKQNLERFKKGDVRFLICTDVAARGIDIHGVPYVINVTLPDEKQNYVHRIGRVGRAERMGLAISLVATEKEKVWYHVCSSRGKGCYNTRLKEDGGCTIWYNEMQLLSEIEEHLNCTISQVEPDIKVPVDEFDGKVTYGQKRAAGGGSYKGHVDILAPTVQELAALEKEAQTSFLHLGYLPNQLFRTF | |
RUO | |
DDX1 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
1653 | |
DDX1 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DBP-RB/UKVH5d | |
DDX1 | |
Recombinant | |
wheat germ expression system |