missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human DCTD Partial ORF (NP_001912.2, 69 a.a. - 178 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00001635-Q01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16107064

  • 349.00€ / 10µg

Estimated Shipment: 04-07-2023
to see stock.
Add to basket

Description

Description

The protein encoded by this gene catalyzes the deamination of dCMP to dUMP, the nucleotide substrate for thymidylate synthase. The encoded protein is allosterically activated by dCTP and inhibited by dTTP, and is found as a homohexamer. This protein uses zinc as a cofactor for its activity. Two transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq]

Sequence: RTAENKLDTKYPYVCHAELNAIMNKNSTDVKGCSMYVALFPCNECAKLIIQAGIKEVIFMSDKYHDSDEATAARLLFNMAGVTFRKFIPKCSKIVIDFDSINSRPSQKLQ
Specifications

Specifications

NP_001912.2
Liquid
1635
DCTD (Human) Recombinant Protein (Q01)
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
MGC111062
DCTD
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
37.84kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
RTAENKLDTKYPYVCHAELNAIMNKNSTDVKGCSMYVALFPCNECAKLIIQAGIKEVIFMSDKYHDSDEATAARLLFNMAGVTFRKFIPKCSKIVIDFDSINSRPSQKLQ
RUO
DCTD
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers