missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-SIGN/CD209 Antibody, R&D Systems™

Description
DC-SIGN/CD209 Monoclonal specifically detects DC-SIGN/CD209 in Human samples. It is validated for Western Blot, Flow Cytometry, Immunohistochemistry, Adhesion Blockade, Immunocytochemistry, CyTOF-ready.
Specifications
Specifications
| Antigen | DC-SIGN/CD209 |
| Applications | Western Blot, Flow Cytometry, Adhesion Blockade, CyTOF, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | 120507 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, Flow Cytometry 2.5 ug/10^6 cells, Adhesion Blockade 5.0 ug/mL, CyTOF-ready |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Alias | CD209, CD209 antigendendritic cell-specific intracellular adhesion molecules (ICAM)-3 grabbingnon-integrin, CD209 molecule, CDSIGNHIV gpl20-binding protein, CLEC4L, CLEC4LC-type lectin domain family 4 member L, DCSIGN, DC-SIGN1, DC-SIGN1C-type lectin domain family 4, member L, DC-SIGNMGC129965, Dendritic cell-specific ICAM-3-grabbing non-integrin 1 |
| Gene Symbols | CD209 |
| Host Species | Mouse |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?