missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DC-LAMP Antibody, R&D Systems™

Description
DC-LAMP Monoclonal specifically detects DC-LAMP in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | DC-LAMP |
| Applications | Western Blot |
| Classification | Monoclonal |
| Clone | 530217 |
| Concentration | LYOPH |
| Conjugate | Unconjugated |
| Dilution | Western Blot 2 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | EAW78338 |
| Gene Alias | CD208, CD208 antigen, DCLAMP, DC-LAMP, DCLAMPDC-lysosome-associated membrane glycoprotein, LAMP, LAMP3, LAMP-3, lysosomal-associated membrane protein 3DC LAMP, lysosome-associated membrane glycoprotein 3, Protein TSC403, TSC403, TSC403CD208 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?