missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human DAN Antibody, R&D Systems™

Description
DAN/NBL1 Monoclonal specifically detects DAN/NBL1 in Human samples. It is validated for Western Blot, ELISA Capture (Matched Antibody Pair).
Specifications
Specifications
| Antigen | DAN/NBL1 |
| Applications | Western Blot, ELISA |
| Classification | Monoclonal |
| Clone | 161502 |
| Conjugate | Unconjugated |
| Dilution | Western Blot 1 ug/mL, ELISA Capture (Matched Antibody Pair) 2-8 ug/mL |
| Formulation | Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative |
| Gene Accession No. | P41271 |
| Gene Alias | DAN domain family member 1, DAND1, MGC8972, NBL1, neuroblastoma suppressor of tumorigenicity 1, neuroblastoma, suppression of tumorigenicity 1, Protein N03, suppression of tumorigenicity 1, Zinc finger protein DAN |
| Gene Symbols | NBL1 |
| Show More |
For Research Use Only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?