Learn More
Human Cystatin SA Antibody, R&D Systems™
GREENER_CHOICE
Mouse Monoclonal Antibody
Brand:  R&D Systems MAB1201-SP
 View more versions of this product
Product Code. 18753749
  • 184.00€ / 25µg
Description
Cystatin SA Monoclonal specifically detects Cystatin SA in Human samples. It is validated for Western Blot.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Cystatin SA
Monoclonal
LYOPH
Western Blot 1 ug/mL
P09228
CST2
Mouse myeloma cell line NS0-derived recombinant human Cystatin SA aa 21-141 Accession # P09228
25 μg
Primary
Detects human Cystatin SA in direct ELISAs and Western blots. In direct ELISAs, this antibody shows 100% cross-reactivity with recombinant human (rh) Cystatin S and SN and no cross-reactivity with rhCystatin A, B, C, D, E/M, rhFetuin A, B, and rhHPRG (Histidine-Proline-Rich Glycoprotein).
Human
Purified
Western Blot
202332
Unconjugated
Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative
CST2, CSTSA, cystatin 2, cystatin S5, cystatin SA, Cystatin-2, Cystatin-S5, cystatin-SA, cysteine-proteinase inhibitor, MGC71924, salivary cysteine (thiol) protease inhibitor
Mouse
Protein A or G purified from hybridoma culture supernatant
RUO
1470
Reconstitute at 0.5 mg/mL in sterile PBS.
Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution.
IgG2b