missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Human Cystatin B Antibody, R&D Systems™

Mouse Monoclonal Antibody has been used in 2 publications
Brand: R&D Systems MAB1408-SP
This item is not returnable.
View return policy
Description
Cystatin B/Stefin B Monoclonal specifically detects Cystatin B/Stefin B in Human samples. It is validated for Western Blot, Immunoprecipitation, Neutralization, Knockout Validated.
Specifications
| Cystatin B/Stefin B | |
| Monoclonal | |
| Unconjugated | |
| Lyophilized from a 0.2 μm filtered solution in PBS with Trehalose. *Small pack size (SP) is supplied as a 0.2 μm filtered solution in PBS. with No Preservative | |
| CPI-B, CST6cystatin B (liver thiol proteinase inhibitor)10STFBcystatin-B, CSTB, cystatin B (stefin B), EPM1, Liver thiol proteinase inhibitor, PME, stefin B, stefin-B | |
| Mouse | |
| Protein A or G purified from hybridoma culture supernatant | |
| RUO | |
| 1476 | |
| Reconstitute at 0.5 mg/mL in sterile PBS. | |
| Use a manual defrost freezer and avoid repeated freeze-thaw cycles. 12 months from date of receipt, -20 to -70 degreesC as supplied. 1 month, 2 to 8 degreesC under sterile conditions after reconstitution. 6 months, -20 to -70 degreesC under sterile conditions after reconstitution. | |
| IgG2b |
| Western Blot, Immunoprecipitation, Neutralization, Immunoassay | |
| 225228 | |
| Western Blot 2 ug/mL, Immunoprecipitation 25 ug/mL, Neutralization 5.4 ug/mL, Knockout Validated | |
| P04080 | |
| CSTB | |
| E. coli-derived recombinant human Cystatin B aa Met2-Phe98 Accession # P04080 | |
| 25 μg | |
| Primary | |
| Detects human Cystatin B in Western blots and direct ELISAs. In Western blots, less than 5% cross-reactivity with recombinant human (rh) Cystatins A and S, recombinant mouse Cystatin B, rhFetuin A, and rhHPRG (Histidine-Proline-Rich Glycoprotein) is observed and approximately 5% to 10% cross-reactivity with rhCystatins C, D, E/M, SA, SN, and rhFetuin B is observed. | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction