Learn More
Abnova™ Human CYP3A43 Full-length ORF (AAI00983.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
Brand: Abnova™ H00064816-P01.10ug
Additional Details : Weight : 0.00010kg
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme has a low level of testosterone hydroxylase activity. Although it bears homology to some drug-metabolizing cytochrome P450s, it is unknown whether the enzyme is also involved in xenobiotic metabolism. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Alternate splicing of this gene results in three transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MNLETGNGPHHFPMWRYVGEKPEAGSREQQVHQLERFLWGLHHGCNHWHIIWSELGFSQQSTRSLSEKYEEAFKIGFFGSLFTLNIQSLAVLPRPECMVRSGLTETSTSWVRVILLPQPPEELGLQHRVDFFQQIIDSQNSKETKSHKALSDLELVAQSIIIIFAAYDTTSTTLPFIMYELATHPDVQQKLQEEIDAVLPNKAPVTYDALVQMEYLDMVVNETLRLFPVVSRVTRVCKKDIEINGVFIPKGLAVMVPIYALHHDPKYWTEPEKFCPERSHSpecifications
AAI00983.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
10 ug | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119315/MGC119316 | |
CYP3A43 | |
Recombinant | |
wheat germ expression system |
Antibody Production, Protein Array, ELISA, Western Blot | |
64816 | |
CYP3A43 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
MNLETGNGPHHFPMWRYVGEKPEAGSREQQVHQLERFLWGLHHGCNHWHIIWSELGFSQQSTRSLSEKYEEAFKIGFFGSLFTLNIQSLAVLPRPECMVRSGLTETSTSWVRVILLPQPPEELGLQHRVDFFQQIIDSQNSKETKSHKALSDLELVAQSIIIIFAAYDTTSTTLPFIMYELATHPDVQQKLQEEIDAVLPNKAPVTYDALVQMEYLDMVVNETLRLFPVVSRVTRVCKKDIEINGVFIPKGLAVMVPIYALHHDPKYWTEPEKFCPERSH | |
RUO | |
CYP3A43 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |