Learn More
Abnova™ Human CYP3A43 Full-length ORF (AAI00983.1, 1 a.a. - 280 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAI00983.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 64816 |
Molecular Weight (g/mol) | 58.6kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16153763
|
Abnova™
H00064816-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
16163763
|
Abnova™
H00064816-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 05-06-2024
Log in to see stock. |
|||||
Description
This gene encodes a member of the cytochrome P450 superfamily of enzymes. The cytochrome P450 proteins are monooxygenases which catalyze many reactions involved in drug metabolism and synthesis of cholesterol, steroids and other lipids. This enzyme has a low level of testosterone hydroxylase activity. Although it bears homology to some drug-metabolizing cytochrome P450s, it is unknown whether the enzyme is also involved in xenobiotic metabolism. This gene is part of a cluster of cytochrome P450 genes on chromosome 7q21.1. Alternate splicing of this gene results in three transcript variants encoding different isoforms. [provided by RefSeq]
Sequence: MNLETGNGPHHFPMWRYVGEKPEAGSREQQVHQLERFLWGLHHGCNHWHIIWSELGFSQQSTRSLSEKYEEAFKIGFFGSLFTLNIQSLAVLPRPECMVRSGLTETSTSWVRVILLPQPPEELGLQHRVDFFQQIIDSQNSKETKSHKALSDLELVAQSIIIIFAAYDTTSTTLPFIMYELATHPDVQQKLQEEIDAVLPNKAPVTYDALVQMEYLDMVVNETLRLFPVVSRVTRVCKKDIEINGVFIPKGLAVMVPIYALHHDPKYWTEPEKFCPERSHSpecifications
AAI00983.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
58.6kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNLETGNGPHHFPMWRYVGEKPEAGSREQQVHQLERFLWGLHHGCNHWHIIWSELGFSQQSTRSLSEKYEEAFKIGFFGSLFTLNIQSLAVLPRPECMVRSGLTETSTSWVRVILLPQPPEELGLQHRVDFFQQIIDSQNSKETKSHKALSDLELVAQSIIIIFAAYDTTSTTLPFIMYELATHPDVQQKLQEEIDAVLPNKAPVTYDALVQMEYLDMVVNETLRLFPVVSRVTRVCKKDIEINGVFIPKGLAVMVPIYALHHDPKYWTEPEKFCPERSH | |
RUO | |
CYP3A43 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
64816 | |
CYP3A43 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
MGC119315/MGC119316 | |
CYP3A43 | |
Recombinant | |
wheat germ expression system |