missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human COG8 Full-length ORF (AAH17492.1, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00084342-P01.10ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16195853

  • 349.00€ / 10µg

Estimated Shipment: 31-10-2023
to see stock.
Add to basket

Description

Description

This gene encodes a protein that is a component of the conserved oligomeric Golgi (COG) complex, a multiprotein complex that plays a structural role in the Golgi apparatus, and is involved in intracellular membrane trafficking and glycoprotein modification. Mutations in this gene cause congenital disorder of glycosylation, type IIh, a disease that is characterized by under-glycosylated serum proteins, and whose symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance. [provided by RefSeq]

Sequence: MNSYMLISAPAILGTSNMPAAVPATQPGTLQPPMVLLDFPPLACFLNNILVAFNDLRLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNIGAIQEPLAFILPKRETLFTLDDQALGPELTAPAPEPPAEEPRLEPAGPACPEGGRAETQAEPPSVGP
Specifications

Specifications

AAH17492.1
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
49.7kDa
Glutathione Sepharose 4 Fast Flow
10 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
DOR1/FLJ22315
COG8
Recombinant
wheat germ expression system
Antibody Production, Protein Array, ELISA, Western Blot
84342
COG8 (Human) Recombinant Protein (P01)
12.5% SDS-PAGE Stained with Coomassie Blue.
MNSYMLISAPAILGTSNMPAAVPATQPGTLQPPMVLLDFPPLACFLNNILVAFNDLRLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNIGAIQEPLAFILPKRETLFTLDDQALGPELTAPAPEPPAEEPRLEPAGPACPEGGRAETQAEPPSVGP
RUO
COG8
Wheat Germ (in vitro)
GST
Liquid
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers