Learn More
Abnova™ Human COG8 Full-length ORF (AAH17492.1, 1 a.a. - 219 a.a.) Recombinant Protein with GST-tag at N-terminal
Used for AP, Array, ELISA, WB-Re
335.00€ - 508.00€
Specifications
Accession Number | AAH17492.1 |
---|---|
For Use With (Application) | Antibody Production, Protein Array, ELISA, Western Blot |
Formulation | 50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. |
Gene ID (Entrez) | 84342 |
Molecular Weight (g/mol) | 49.7kDa |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16105863
|
Abnova™
H00084342-P01.25UG |
25 ug |
508.00€
25µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
16195853
|
Abnova™
H00084342-P01.10UG |
10 ug |
335.00€
10µg |
Estimated Shipment: 07-05-2024
Log in to see stock. |
|||||
Description
This gene encodes a protein that is a component of the conserved oligomeric Golgi (COG) complex, a multiprotein complex that plays a structural role in the Golgi apparatus, and is involved in intracellular membrane trafficking and glycoprotein modification. Mutations in this gene cause congenital disorder of glycosylation, type IIh, a disease that is characterized by under-glycosylated serum proteins, and whose symptoms include severe psychomotor retardation, failure to thrive, seizures, and dairy and wheat product intolerance. [provided by RefSeq]
Sequence: MNSYMLISAPAILGTSNMPAAVPATQPGTLQPPMVLLDFPPLACFLNNILVAFNDLRLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNIGAIQEPLAFILPKRETLFTLDDQALGPELTAPAPEPPAEEPRLEPAGPACPEGGRAETQAEPPSVGPSpecifications
AAH17492.1 | |
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer. | |
49.7kDa | |
Glutathione Sepharose 4 Fast Flow | |
MNSYMLISAPAILGTSNMPAAVPATQPGTLQPPMVLLDFPPLACFLNNILVAFNDLRLCCPVALAQDVTGALEDALAKVTKIILAFHRAEEAAFSSGEQELFVQFCTVFLEDLVPYLNRCLQVLFPPAQIAQTLGIPPTQLSKYGNLGHVNIGAIQEPLAFILPKRETLFTLDDQALGPELTAPAPEPPAEEPRLEPAGPACPEGGRAETQAEPPSVGP | |
RUO | |
COG8 | |
Wheat Germ (in vitro) | |
GST | |
Liquid |
Antibody Production, Protein Array, ELISA, Western Blot | |
84342 | |
COG8 (Human) Recombinant Protein (P01) | |
12.5% SDS-PAGE Stained with Coomassie Blue. | |
Store at -80°C. Aliquot to avoid repeated freezing and thawing. | |
DOR1/FLJ22315 | |
COG8 | |
Recombinant | |
wheat germ expression system |