missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ Human CNOT2 Partial ORF (NP_055330, 441 a.a. - 540 a.a.) Recombinant Protein with GST-tag at N-terminal

Used for AP, Array, ELISA, WB-Re

Brand:  Abnova™ H00004848-Q01.25ug

Additional Details : Weight : 0.00010kg

 View more versions of this product

Product Code. 16102015

  • 508.00€ / 25µg

Estimated Shipment: 12-05-2023
to see stock.
Add to basket

Description

Description

Sequence: EDLLFYLYYMNGGDVLQLLAAVELFNRDWRYHKEERVWITRAPGMEPTMKTNTYERGTYYFFDCLNWRKVAKEFHLEYDKLEERPHLPSTFNYNPAQQAF
Specifications

Specifications

NP_055330
Liquid
4848
CNOT2 (Human) Recombinant Protein (Q01)
25 ug
Store at -80°C. Aliquot to avoid repeated freezing and thawing.
CDC36/HSPC131/NOT2/NOT2H
CNOT2
Yes
wheat germ expression system
Antibody Production, Enzyme-linked Immunoabsorbent Assay, Protein Array, Western Blot (Recombinant protein)
50mM Tris-HCI, 10mM reduced Glutathione, pH=8.0 in the elution buffer.
36.74kDa
12.5% SDS-PAGE Stained with Coomassie Blue.
EDLLFYLYYMNGGDVLQLLAAVELFNRDWRYHKEERVWITRAPGMEPTMKTNTYERGTYYFFDCLNWRKVAKEFHLEYDKLEERPHLPSTFNYNPAQQAF
RUO
CNOT2
Wheat Germ (in vitro)
GST
Videos
SDS
Documents

Documents

Certificates
Special Offers

Special Offers